5-Amino-4-bromo-2-fluorobenzonitrile 96%

Supplier: APOLLO SCIENTIFIC

PC904887-10G PC904887-1G PC904887-5G
77418-522EA 637.78 USD
77418-522 77418-524 77418-526
5-Amino-4-bromo-2-fluorobenzonitrile 96%
5-Amino-4-bromo-2-fluorobenzonitrile
Formula: C₇H₄BrFN₂
Storage Temperature: Ambient
MDL Number: MFCD11977386
CAS Number: 893615-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR