2-(Trifluoromethyl)cyclohexanone 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC7668L-25G PC7668L-5G
77418-098EA 1301.02 USD
77418-098 77418-100
2-(Trifluoromethyl)cyclohexanone 97%
2-(Trifluoromethyl)cyclohexanone
Formula: C₇H₉F₃O
MW: 166.14 g/mol
Boiling Pt: 71…73 °C/12 mmHg
Storage Temperature: Ambient
MDL Number: MFCD00102125
CAS Number: 56734-74-8
UN: 1224
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR