Tetramethylammonium tetrafluoroborate 97%

Supplier: APOLLO SCIENTIFIC

PC6938-25G PC6938-5G
77417-610EA 100.86 USD
77417-610 77417-612
Tetramethylammonium tetrafluoroborate 97%
Tetramethylammonium tetrafluoroborate
Formula: C₄H₁₂BF₄N
Melting Pt: 417…419 °C
Storage Temperature: Ambient
MDL Number: MFCD00011745
CAS Number: 661-36-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR