2,3,5,6-Tetrafluorotoluene 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC6930-10G PC6930-1G PC6930-5G
77417-598EA 410.58 USD
77417-598 77417-600 77417-602
2,3,5,6-Tetrafluorotoluene 98%
2,3,5,6-Tetrafluorotoluene
Formula: C₇H₄F₄
MW: 164.1 g/mol
Boiling Pt: 125…126 °C
Density: 1.347 g/cm³ (25 °C)
Flash Pt: 25 °C
Storage Temperature: Ambient
CAS Number: 5230-78-4
EINECS: 226-028-5
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR