Perfluorocyclohexanecarbonyl fluoride 65%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Undecafluorocyclohexanecarbonyl fluoride, (Fluorocarbonyl)perfluorocyclohexane

PC5932E-10G PC5932E-50G
77417-188EA 21.26 USD
77417-188 77417-190
Perfluorocyclohexanecarbonyl fluoride 65%
Perfluorocyclohexanecarbonyl fluoride
Formula: C₇F₁₂O
MW: 328.055 g/mol
Boiling Pt: 70 °C
Density: 1.6 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00236697
CAS Number: 6588-63-2
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR