3-Fluoro-5-methylphenylacetonitrile 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC5919-10G PC5919-1G PC5919-5G
77417-176EA 275.08 USD
77417-176 77417-178 77417-180
3-Fluoro-5-methylphenylacetonitrile 97%
3-Fluoro-5-methylphenylacetonitrile
Formula: C₉H₈FN
Storage Temperature: Ambient
MDL Number: MFCD03094317
CAS Number: 518070-21-8
UN: 3276
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR