4-Chloro-2,6-difluorophenylacetonitrile 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC8244-1G
PC8244-5G
PC8244-10G
77418-374EA
95.76
USD
77418-374
77418-376
77418-372
4-Chloro-2,6-difluorophenylacetonitrile 98%
4-Chloro-2,6-difluorophenylacetonitrile
Formula:
C₈H₄ClF₂N Storage Temperature: Ambient |
MDL Number:
MFCD03094465 CAS Number: 537033-53-7 UN: 3439 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...