4,4-Difluorocyclohexanone

Supplier: APOLLO SCIENTIFIC
Danger

PC7922-10G PC7922-1G PC7922-5G
77418-302EA 108 USD
77418-302 77418-304 77418-306
4,4-Difluorocyclohexanone
4,4-Difluorocyclohexanone
Formula: C₆H₈F₂O
Melting Pt: 34…37 °C
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD08146635
CAS Number: 22515-18-0
UN: 1325
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR