1,3,5-Tris(trifluoromethyl)benzene 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC7919-5G PC7919-25G
77418-294EA 99.84 USD
77418-294 77418-292
1,3,5-Tris(trifluoromethyl)benzene 97%
1,3,5-Tris(trifluoromethyl)benzene
Formula: C₉H₃F₉
MW: 282.105 g/mol
Boiling Pt: 119.6 °C/750 mmHg
Melting Pt: 9 °C
Density: 1.5139 g/cm³ (25 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00054722
CAS Number: 729-81-7
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR