3,3,3-Trifluoro-2-hydroxy-2-methylpropionic acid 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3,3,3-Trifluoro-2-hydroxyisobutyric Acid, 3,3,3-Trifluoro-2-hydroxy-2-methylpropanoic acid, 2-Hydroxy-2-(trifluoromethyl)propionic Acid

PC7674-5G PC7674-1G PC7674-25G
77418-114EA 87.62 USD
77418-114 77418-110 77418-112
3,3,3-Trifluoro-2-hydroxy-2-methylpropionic acid 97%
3,3,3-Trifluoro-2-hydroxy-2-methylpropionic acid
Formula: C₄H₅F₃O₃
MW: 158.08 g/mol
Boiling Pt: 189 °C
Melting Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00190646
CAS Number: 374-35-6
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR