3-(Trifluoromethyl)benzylamine 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC7629-100G PC7629-25G PC7629-5G
77418-034EA 151.8 USD
77418-034 77418-036 77418-038
3-(Trifluoromethyl)benzylamine 97%
3-(Trifluoromethyl)benzylamine
Formula: C₈H₈F₃N
Boiling Pt: 93…97 °C/28 mmHg
Density: 1.222 g/cm³ (25 °C)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00008117
CAS Number: 2740-83-2
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR