3-(Trifluoromethoxy)benzonitrile 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC7438F-10G PC7438F-25G PC7438F-5G
77417-884EA 23.44 USD
77417-884 77417-886 77417-888
3-(Trifluoromethoxy)benzonitrile 97%
3-(Trifluoromethoxy)benzonitrile
Formula: C₈H₄F₃NO
Density: 1.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00040959
CAS Number: 52771-22-9
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR