5,5,5-Trifluoropentanonitrile 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC7318-1G PC7318-5G
77417-810EA 63.16 USD
77417-810 77417-812
5,5,5-Trifluoropentanonitrile 98%
5,5,5-Trifluoropentanonitrile
Formula: C₅H₆F₃N
Boiling Pt: 60 °C (10 mmHg)
Storage Temperature: Refrigerator
MDL Number: MFCD08059547
CAS Number: 89866-61-5
UN: 3275
ADR: 6.1+3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR