2,3,5,6-Tetrafluorophenol 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC6820-25G PC6820-50G
77417-542EA 44.82 USD
77417-542 77417-544
2,3,5,6-Tetrafluorophenol 98%
2,3,5,6-Tetrafluorophenol
Formula: C₆H₂F₄O
Boiling Pt: 140…143 °C
Melting Pt: 33…39 °C
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00002157
CAS Number: 769-39-1
EINECS: 212-209-6
UN: 2923
ADR: 8+6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR