2,2-Bis(trifluoromethyl)-1,3-dioxolane 99%

Supplier: APOLLO SCIENTIFIC

Synonyms: 1,1,1,3,3,3-Hexafluoropropan-2-one ethylene ketal

PC9861-1G PC9861-250MG PC9861-5G
77418-736EA 269.98 USD
77418-736 77418-738 77418-740
2,2-Bis(trifluoromethyl)-1,3-dioxolane 99%
2,2-Bis(trifluoromethyl)-1,3-dioxolane
Formula: C₅H₄F₆O₂
MW: 210.074 g/mol
Boiling Pt: 106…107 °C
Density: 1.53 g/cm³ (25 °C)
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00191832
CAS Number: 1765-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR