(S)-2-(Trifluoromethyl)pyrrolidine 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC8535-1G PC8535-25G PC8535-5G
77418-424EA 29.54 USD
77418-424 77418-426 77418-428
(S)-2-(Trifluoromethyl)pyrrolidine 97%
(S)-2-(Trifluoromethyl)pyrrolidine
Formula: C₅H₈F₃N
MW: 139.12 g/mol
Boiling Pt: 110…112 °C
Melting Pt: 27…33 °C
Flash Pt: 26 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03093766
CAS Number: 119580-41-5
UN: 1325
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR