2-(Ethylamino)-6-(trifluoromethyl)-4-pyrimidinol

Supplier: APOLLO SCIENTIFIC
Warning

PC8467-1G PC8467-5G
77418-414EA 135.5 USD
77418-414 77418-416
2-(Ethylamino)-6-(trifluoromethyl)-4-pyrimidinol
2-(Ethylamino)-6-(trifluoromethyl)-4-pyrimidinol
Formula: C₇H₈F₃N₃O
Melting Pt: 198…200 °C
Storage Temperature: Ambient
MDL Number: MFCD00171969
CAS Number: 338792-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR