2-Bromo-2'-(trifluoromethoxy)acetophenone 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-(Trifluoromethoxy)phenacyl bromide

PC7361-1G PC7361-25G PC7361-5G
77417-832EA 93.4 USD
77417-832 77417-834 77417-836
2-Bromo-2'-(trifluoromethoxy)acetophenone 97%
2-Bromo-2'-(trifluoromethoxy)acetophenone
Formula: C₉H₆BrF₃O₂
Boiling Pt: 52 °C/0.04 mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD03094424
CAS Number: 530141-40-3
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR