2,4-Dimethoxy-5-fluoroaniline

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 5-Fluoro-2,4-dimethoxyaniline

PC7349-1G PC7349-250MG PC7349-5G
77417-824EA 96.78 USD
77417-824 77417-826 77417-828
2,4-Dimethoxy-5-fluoroaniline
2,4-Dimethoxy-5-fluoroaniline
Formula: C₈H₁₀FNO₂
MW: 171.17 g/mol
Melting Pt: 47…49 °C
Storage Temperature: Ambient
MDL Number: MFCD12547861
CAS Number: 195136-65-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR