2-Fluoro-6-methoxybenzamide

Supplier: APOLLO SCIENTIFIC
Warning

PC7323-1G PC7323-5G
77417-818EA 144.68 USD
77417-818 77417-820
2-Fluoro-6-methoxybenzamide
2-Fluoro-6-methoxybenzamide
Formula: C₈H₈FNO₂
Storage Temperature: Ambient
MDL Number: MFCD00277474
CAS Number: 529512-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR