2,4,5-Trifluorobenzyl bromide 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-(Bromomethyl)-2,4,5-trifluorobenzene, α-Bromo-2,4,5-trifluorotoluene

PC7285S-100G PC7285S-25G PC7285S-500G
77417-738EA 192.56 USD
77417-738 77417-740 77417-742
2,4,5-Trifluorobenzyl bromide 97%
2,4,5-Trifluorobenzyl bromide
Formula: C₇H₄BrF₃
MW: 225.01 g/mol
Density: 1.71 g/cm³ (25 °C)
Flash Pt: 90.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00061209
CAS Number: 157911-56-3
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR