5-Amino-3-fluorobenzonitrile 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-Amino-5-fluorobenzonitrile, 3-Cyano-5-fluoroaniline

PC7168-25G PC7168-5G
77417-672EA 190.52 USD
77417-672 77417-674
5-Amino-3-fluorobenzonitrile 97%
5-Amino-3-fluorobenzonitrile
Formula: C₇H₅FN₂
MW: 136.13 g/mol
Melting Pt: 105…107 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD05864357
CAS Number: 210992-28-2
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR