Trifluoroacetic acid 99,5%

Supplier: APOLLO SCIENTIFIC
Danger

PC7160A-1L PC7160A-500ML
77417-664EA 88.64 USD
77417-664 77417-666
Trifluoroacetic acid 99,5%
Trifluoroacetic acid

Plastic coated bottle with tamper evident cap

Formula: C₂HF₃O₂
Boiling Pt: 72.4 °C
Melting Pt: –15.4 °C
Density: 1.48 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR