1H,1H,12H,12H-Eicosafluoro-1,12-dodecandiol 96%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1H,1H,12H,12H-Perfluoro-1,12-dodecanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Eicosafluoro-1,12-dodecandiol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-Icosafluorododecane-1,12-diol, 1H,1H,12H,12H-Icosafluoro-1,12-dodecanediol, 1H,1H,12H,12H-Perfluoro-1,12-dodecandiol

PC5993-1G PC5993-25G PC5993-5G
77417-236EA 63.16 USD
77417-236 77417-238 77417-240
1H,1H,12H,12H-Eicosafluoro-1,12-dodecandiol 96%
1H,1H,12H,12H-Eicosafluoro-1,12-dodecandiol
Formula: C₁₂H₆F₂₀O₂
MW: 562.15 g/mol
Boiling Pt: 170 °C/5 mmHg
Melting Pt: 156…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00236625
CAS Number: 183162-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR