2,3,5-Trifluoroaniline 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC5403-1G PC5403-25G PC5403-5G
77416-726EA 21.32 USD
77416-726 77416-728 77416-730
2,3,5-Trifluoroaniline 98%
2,3,5-Trifluoroaniline
Formula: C₆H₄F₃N
MW: 147.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00083549
CAS Number: 363-80-4
UN: 2941
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR