5,5,5-Trifluoro-DL-leucine 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC7392-100MG PC7392-1G PC7392-250MG
77417-854EA 151.8 USD
77417-854 77417-856 77417-858
5,5,5-Trifluoro-DL-leucine 97%
5,5,5-Trifluoro-DL-leucine
Formula: C₆H₁₀F₃NO₂
MW: 185.15 g/mol
Melting Pt: 272…275 °C
Storage Temperature: Ambient
MDL Number: MFCD00039528
CAS Number: 2792-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR