1,3,5-Trifluorobenzene 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC7280-100G PC7280-25G PC7280-5G
77417-702EA 123.28 USD
77417-702 77417-704 77417-706
1,3,5-Trifluorobenzene 98%
1,3,5-Trifluorobenzene
Formula: C₆H₃F₃
MW: 132.09 g/mol
Boiling Pt: 76 °C
Melting Pt: –5.5 °C
Density: 1.277 g/cm³ (25 °C)
Flash Pt: –7 °C
Storage Temperature: Ambient
MDL Number: MFCD00000333
CAS Number: 372-38-3
EINECS: 206-751-2
UN: 2387
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR