1,1,1,2,2-Pentafluoro-3-methylbutan-3-ol

Supplier: APOLLO SCIENTIFIC
Warning

PC7242-1G PC7242-250MG PC7242-5G
77417-694EA 339.26 USD
77417-694 77417-696 77417-698
1,1,1,2,2-Pentafluoro-3-methylbutan-3-ol
1,1,1,2,2-Pentafluoro-3-methylbutan-3-ol
Formula: C₅H₇F₅O
MW: 178.1 g/mol
Boiling Pt: 90…92 °C
Density: 1.24 g/cm³ (25 °C)
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00069121
CAS Number: 374-46-9
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR