3-Bromo-5-(trifluoromethyl)benzenesulfonyl chloride 97%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC6577-1G
PC6577-25G
PC6577-5G
77417-448EA
21.26
USD
77417-448
77417-450
77417-452
3-Bromo-5-(trifluoromethyl)benzenesulfonyl chloride 97%
3-Bromo-5-(trifluoromethyl)benzenesulphonyl chloride
Formula:
C₇H₃BrClF₃O₂S Boiling Pt: 217…218 °C Density: 1.835 g/cm³ (25 °C) Flash Pt: 66 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094393 CAS Number: 351003-46-8 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...