3-Fluoro-4-methylbenzenesulfonyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-Fluoro-4-methylbenzene-1-sulphonyl chloride

PC6569-25G PC6569-5G
77417-440EA 275.08 USD
77417-440 77417-442
3-Fluoro-4-methylbenzenesulfonyl chloride 97%
3-Fluoro-4-methylbenzenesulphonyl chloride
Formula: C₇H₆ClFO₂S
MW: 208.64 g/mol
Melting Pt: 46…50 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03094389
CAS Number: 90260-13-2
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR