N-Pentafluorophenyldichloromaleimide 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC5663E-1G PC5663E-5G
77416-856EA 89.36 USD
77416-856 77416-858
N-Pentafluorophenyldichloromaleimide 97%
N-Pentafluorophenyldichloromaleimide
Formula: C₁₀Cl₂F₅NO₂
Melting Pt: 148…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00054728
CAS Number: 186958-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR