Methyl 2,2,2-trifluoroethyl carbonate 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: CArbonic acid, methyl 2,2,2-trifluoroethyl ester, Methyl 2,2,2-trifluoroethyl ester, Carbonic Acid

PC53009-1G PC53009-25G PC53009-5G
77416-602EA 21.26 USD
77416-602 77416-604 77416-606
Methyl 2,2,2-trifluoroethyl carbonate 95%
Methyl 2,2,2-trifluoroethyl carbonate
Formula: C₄H₅F₃O₃
MW: 158.076 g/mol
Boiling Pt: 57…59 °C ( 120 mmHg)
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD28054295
CAS Number: 156783-95-8
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR