Methyl-3-(2-fluoro-4-nitrophenoxy)-2-thiophenecarboxylate 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC9409-100MG PC9409-1G
77418-674EA 161.71 USD
77418-674 77418-670 77418-672
Methyl-3-(2-fluoro-4-nitrophenoxy)-2-thiophenecarboxylate 97%
Methyl-3-(2-fluoro-4-nitrophenoxy)-2-thiophenecarboxylate
Formula: C₁₂H₈FNO₅S
Melting Pt: 112…114 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00202715
CAS Number: 252026-46-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR