2-Fluoro-6-iodobenzaldehyde 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 6-Fluoro-2-Iodobenzaldehyde

PC9246-1G PC9246-25G PC9246-5G
77418-624EA 21.36 USD
77418-624 77418-626 77418-628
2-Fluoro-6-iodobenzaldehyde 97%
2-Fluoro-6-iodobenzaldehyde
Formula: C₇H₄FIO
Melting Pt: 36…40 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD07371506
CAS Number: 146137-72-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR