5-Amino-2-(trifluoromethyl)benzimidazole 96%

Supplier: APOLLO SCIENTIFIC
Danger

PC9213-1G PC9213-5G
77418-616EA 50.94 USD
77418-616 77418-618
5-Amino-2-(trifluoromethyl)benzimidazole 96%
5-Amino-2-(trifluoromethyl)benzimidazole
Formula: C₈H₆F₃N₃
MW: 201.15
Melting Pt: 185…189 °C
Storage Temperature: Ambient
MDL Number: MFCD01632182
CAS Number: 3671-66-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR