3-Methoxy-2,4,5-trifluorobenzoyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-Methoxy-2,4,5-trifluorobenzene-1-carbonyl chloride, 2,4,5-Trifluoro-3-methoxybenzoyl chloride

PC7787-100G PC7787-25G PC7787-5G
77418-196EA 108.32 USD
77418-196 77418-198 77418-200
3-Methoxy-2,4,5-trifluorobenzoyl chloride
3-Methoxy-2,4,5-trifluorobenzoyl chloride
Formula: C₈H₄ClF₃O₂
MW: 224.56 g/mol
Boiling Pt: 108…112 °C/20 mmHg
Density: 1.472 g/cm³ (25 °C)
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD02682012
CAS Number: 112811-66-2
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR