Perfluorobutane-1-sulphonyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Nonafluorobutane-1-sulphonyl chloride

PC6996-1G PC6996-25G PC6996-5G
77417-620EA 89.66 USD
77417-620 77417-622 77417-624
Perfluorobutane-1-sulphonyl chloride 97%
Perfluorobutane-1-sulphonyl chloride
Formula: C₄ClF₉O₂S
Boiling Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD03412262
CAS Number: 2991-84-6
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR