Nonafluoro-2-(trifluoromethyl)-2-pentene 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Perfluoro(2-methyl-2-pentene), Perfluoro(2-methylpent-2-ene), 1,1,1,3,4,4,5,5,5-Nonafluoro-2-(trifluoromethyl)-2-pentene

PC6122P-100G PC6122P-1G PC6122P-25G
77417-282EA 251.64 USD
77417-282 77417-284 77417-286
Nonafluoro-2-(trifluoromethyl)-2-pentene 98%
Nonafluoro-2-(trifluoromethyl)-2-pentene
Formula: C₆F₁₂
MW: 300.05 g/mol
Boiling Pt: 48…54 °C
Density: 1.622 g/cm³ (25 °C)
Flash Pt: –12 °C
Storage Temperature: Ambient
MDL Number: MFCD00015724
CAS Number: 1584-03-8
EINECS: 216-436-1
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR