1H,1H,2H-Perfluoro-1-hexene 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC6085D-250G PC6085D-25G PC6085D-100G
77417-274EA 488.02 USD
77417-274 77417-276 77417-272
1H,1H,2H-Perfluoro-1-hexene 98%
1H,1H,2H-Perfluoro-1-hexene
Formula: C₆H₃F₉
Boiling Pt: 59 °C
Density: 1.457 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042338
CAS Number: 19430-93-4
EINECS: 243-053-7
UN: 1993
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR