2,6-Difluoro-4-iodoaniline 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC5886-100G PC5886-25G PC5886-5G
77417-126EA 218.02 USD
77417-126 77417-128 77417-130
2,6-Difluoro-4-iodoaniline 98%
2,6-Difluoro-4-iodoaniline
Formula: C₆H₄F₂IN
MW: 255.0039 g/mol
Melting Pt: 77…79 °C
Storage Temperature: Ambient
MDL Number: MFCD07774189
CAS Number: 141743-49-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR