Methyl-3,3,3-trifluoropropanate

Supplier: APOLLO SCIENTIFIC
Warning

PC5518-1G PC5518-5G
77416-774EA 21.32 USD
77416-774 77416-776
Methyl-3,3,3-trifluoropropanate
Methyl-3,3,3-trifluoropropanate
Formula: C₄H₅F₃O₂
Boiling Pt: 95…96 °C
Storage Temperature: Ambient
MDL Number: MFCD02262206
CAS Number: 18830-44-9
UN: 3272
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR