2-Bromo-3-fluorophenylisothiocyanate

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Bromo-1-fluoro-3-isothiocyanatobenzene

PC502994-1G PC502994-25G PC502994-5G
77416-208EA 75.4 USD
77416-208 77416-210 77416-212
2-Bromo-3-fluorophenylisothiocyanate
2-Bromo-3-fluorophenylisothiocyanate
Formula: C₇H₃BrFNS
MW: 232.073 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22557657
CAS Number: 364364-00-1
UN: 2922
ADR: 8+6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR