1,1-Dichloro-2,2,2-trifluoroethyldifluoromethyl ether 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC6699-25G PC6699-5G
77417-494EA 553.22 USD
77417-494 77417-496
1,1-Dichloro-2,2,2-trifluoroethyldifluoromethyl ether 98%
1,1-Dichloro-2,2,2-trifluoroethyldifluoromethyl ether
Formula: C₃HCl₂F₅O
Boiling Pt: 60…61 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01320775
CAS Number: 32778-07-7
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR