1-[Bis(4-fluorophenyl)methyl]piperazine 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC5636-100G PC5636-25G PC5636-5G
77416-838EA 137.54 USD
77416-838 77416-840 77416-842
1-[Bis(4-fluorophenyl)methyl]piperazine 98%
1-[Bis(4-fluorophenyl)methyl]piperazine
Formula: C₁₇H₁₈F₂N₂
Boiling Pt: 130 °C
Melting Pt: 88…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00038660
CAS Number: 27469-60-9
EINECS: 248-476-0
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR