(2Z)-3-(Diethylamino)-2-fluoroprop-2-enal 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (2Z)-3-(Diethylamino)-2-fluoroacrolein, (2Z)-3-(Diethylamino)-2-fluoroacrylaldehyde

PC53002-1G PC53002-5G
77416-590EA 131.42 USD
77416-590 77416-592
(2Z)-3-(Diethylamino)-2-fluoroprop-2-enal 95%
(2Z)-3-(Diethylamino)-2-fluoroprop-2-enal
Formula: C₇H₁₂FNO
MW: 145.175 g/mol
Storage Temperature: Freezer
MDL Number: MFCD18633001
CAS Number: 152873-64-8
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR