1-(Trifluoromethyl)cyclohexanecarboxylic acid 97%

Supplier: APOLLO SCIENTIFIC

PC520566-1G PC520566-500MG PC520566-5G
77416-500EA 219.04 USD
77416-500 77416-502 77416-504
1-(Trifluoromethyl)cyclohexanecarboxylic acid 97%
1-(Trifluoromethyl)cyclohexanecarboxylic acid
Formula: C₈H₁₁F₃O₂
MW: 196.700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08445820
CAS Number: 180918-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR