Silver bis(trifluoromethanesulfonyl)imide

Supplier: APOLLO SCIENTIFIC

PC50957-25G PC50957-5G
77416-422EA 419.76 USD
77416-422 77416-424
Silver bis(trifluoromethanesulfonyl)imide
Silver bis(trifluoromethanesulfonyl)imide
Formula: C₂AgF₆NO₄S₂
Storage Temperature: Ambient
MDL Number: MFCD08457644
CAS Number: 189114-61-2
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR