5-Bromo-2-fluorophenylisothiocyanate

Supplier: APOLLO SCIENTIFIC
Danger

PC503018-1G PC503018-25G PC503018-5G
77416-346EA 21.4 USD
77416-346 77416-348 77416-350
5-Bromo-2-fluorophenylisothiocyanate
5-Bromo-2-fluorophenylisothiocyanate
Formula: C₇H₃BrFNS
MW: 232.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09800737
CAS Number: 1027513-65-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR