4-Bromo-3-fluorophenyl isothiocyanate

Supplier: APOLLO SCIENTIFIC
Danger

PC503016-1G PC503016-25G PC503016-5G
77416-336EA 45.84 USD
77416-336 77416-338 77416-340
4-Bromo-3-fluorophenyl isothiocyanate
4-Bromo-3-fluorophenyl isothiocyanate
Formula: C₇H₃BrFNS
Storage Temperature: Ambient
CAS Number: 362690-56-0
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR