2-Fluoro-p-toluonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC5912-100G PC5912-25G PC5912-5G
77417-162EA 231.28 USD
77417-162 77417-164 77417-166
2-Fluoro-p-toluonitrile 98%
2-Fluoro-p-toluonitrile
Formula: C₈H₆FN
Melting Pt: 52…53 °C
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD03094311
CAS Number: 85070-67-3
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR